Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat Serpin E2/PN1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB112466 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB112466 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB112466 Supplier Novus Biologicals™ Supplier No. NBP300470
Add to Cart
Add to Cart

Human Serpin E2/PN1 ELISA Kit (Chemiluminescence) measures Serpin E2/PN1 in Serum, plasma, cell supernatant, and other biological fluids.

Specifications

Assay Range 6.25 to 400 pg/mL
Assay Sensitivity 3.75 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Human Serpin E2/PN1 in samples. No significant cross-reactivity or interference between Human Serpin E2/PN1 and analogues was observed.
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 5270
Gene Symbol SERPINE2
Quantity 1 Kit
Research Discipline Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.