Learn More
Invitrogen™ Human Serum Albumin Monoclonal Antibody (10C7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA514717
Description
MA5-14717 targets Human Serum Albumin in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14717 immunogen is human Serum Albumin. MA5-14717 detects Human Serum Albumin which has a predicted molecular weight of approximately 67 kDa. Product MA514717 is a smaller package size of MIH3003 (formerly sold as a Seradyn product).
Albumin is a soluble, monomeric protein with a molecular weight of approximately 65-67 kDa that comprises about one-half of the blood serum protein. Human serum albumin (HAS) is the most abundant protein in human blood plasma, and has a serum half-life of approximately 20 days. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular, unglycosylated serum protein synthesized in the liver as pre-pro-albumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough produce the secreted albumin. Serum albumin has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. The main function of albumin is the regulation of the colloidal osmotic pressure of blood.
Specifications
Human Serum Albumin | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P02768 | |
ALB | |
Human Serum Albumin. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
10C7 | |
Unconjugated | |
ALB | |
Alb; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
Mouse | |
Ion-exchange chromatography | |
RUO | |
213 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.