Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Serum Albumin Native Protein
SDP

Catalog No. NBP196791 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP196791 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196791 Supplier R&D Systems™ Supplier No. NBP196791

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results.

Native Human Serum Albumin. The Human Serum Albumin Native Protein has been validated for the following applications: Western Blot, Fluorophore-linked immunosorbent assay, Immunofluorescence.

Specifications

Accession Number P02768
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10mg/mL Polyethylene Glycol (PEG-8000)
Gene ID (Entrez) 280717
Name Bovine Serum Albumin protein
Quantity 1 mg
Storage Requirements 4°C in the dark.
Cross Reactivity Human
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.