Learn More
Description
Purity: IgG is >98.8% total protein by SDS-PAGE. Becker titer results showed >3.0 mg/mL (antigen/antibody).
Specifications
Specifications
| Antigen | Human Serum Albumin |
| Applications | ELISA, Immunodiffusion, Immunoelectrophoresis, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide; pH 7.4 |
| Gene | ALB |
| Gene Accession No. | P02768 |
| Gene Alias | Alb; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 |
| Gene Symbols | ALB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
