Learn More
Human Serum Amyloid A1/A2 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Serum Amyloid A1/A2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 902770 |
| Conjugate | Unconjugated |
| Dilution | ELISA |
| Formulation | Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | P0DJI8 |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant human Serum Amyloid A1 Arg19-Tyr133 Accession # P0DJI8 |
| Show More |
RUO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.