Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SETDB2 Partial ORF (NP_114121.1, 451 a.a. - 550 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961598 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-598 10 μg
89-961-599 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-598 Supplier Abnova™ Supplier No. H00083852Q02S

Used for AP, Array, ELISA, WB-Re

Proteins that contain a SET domain, such as SETDB2, modulate gene expression epigenetically through histone H3 (see MIM 601128) methylation. SETDB2 is likely a histone H3 methyltransferase, as it contains both the active site and flanking cysteine residues required for catalytic activity (Zhang et al., 2003 [PubMed 12754510]).[supplied by OMIM]

Sequence: HPRTAKTEKCPPKFSNNPKELTMETKYDNISRIQYHSVIRDPESKTAIFQHNGKKMEFVSSESVTPEDNDGFKPPREHLNSKTKGAQKDSSSNHVDEFED

Specifications

Accession Number NP_114121.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 83852
Molecular Weight (g/mol) 36.74kDa
Name SETDB2 (Human) Recombinant Protein (Q02)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen HPRTAKTEKCPPKFSNNPKELTMETKYDNISRIQYHSVIRDPESKTAIFQHNGKKMEFVSSESVTPEDNDGFKPPREHLNSKTKGAQKDSSSNHVDEFED
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C13orf4/CLLD8/CLLL8/DKFZp586I0123/DKFZp761J1217/KMT1F
Common Name SETDB2
Gene Symbol SETDB2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.