Learn More
ACROBiosystems Human SEZ6 25ug
Supplier:  ACROBiosystems SE6H82E325UG
Encompass_Preferred
Biotinylated Human SEZ6 Protein HisAvitag (SE6-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - His 925 (Accession # Q53EL9-1).
Catalog No. 50-253-4186
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.