Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SEZ6L Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5598
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5598 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5598 Supplier R&D Systems Supplier No. AF5598
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SEZ6L Polyclonal specifically detects SEZ6L in Human samples. It is validated for Western Blot.

Specifications

Antigen SEZ6L
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q9BYH1
Gene Alias Acig1, AIG1, Aig1L, BSRP-B, seizure related 6 homolog (mouse)-like, seizure related gene 6-like, SEZ6L
Gene Symbols SEZ6L
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human SEZ6L Leu29-Ala958 Accession # Q9BYH1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23544
Test Specificity Detects human SEZ6L in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse (rm) BSRP-B is observed and less than 2% cross-reactivity with rmBSRP-A and rmBSRP-C is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.