Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SF3B2 Full-length ORF (AAH07610, 1 a.a. - 636 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961618 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-618 25 μg
89-961-617 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-618 Supplier Abnova™ Supplier No. H00010992P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes subunit 2 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex (U2 snRNP). The splicing factor 3b/3a complex binds pre-mRNA upstream of the intron's branch site in a sequence-independent manner and may anchor the U2 snRNP to the pre-mRNA. Splicing factor 3b is also a component of the minor U12-type spliceosome. Subunit 2 associates with pre-mRNA upstream of the branch site at the anchoring site. Subunit 2 also interacts directly with subunit 4 of the splicing factor 3b complex. Subunit 2 is a highly hydrophilic protein with a proline-rich N-terminus and a glutamate-rich stretch in the C-terminus. [provided by RefSeq]

Sequence: MDDPSVGPKIPQALEKILQLKESRQEEMNSQQEEEEMETDARSSLGQSASETEEDTVSVSKKEKNRKRRNRKKKKKPQRVRGVSSESSGDREKDSTRSRGSDSPAADVEIEYVTEEPEIYEPNFIFFKRIFEAFKLTDDVKKEKEKEPEKLDKLENSAAPKKKGFEEEHKDSDDDSSDDEQEKKPEAPKLSKKKLRRMNRFTVAELKQLVARPDVVEMHDVTAQDPKLLVHLKATRNSVPVPRHWCFKRKYLQGKRGIEKPPFELPDFIKRTGIQEMREALQEKEEQKTMKSKMREKVRPKMGKIDIDYQKLHDAFFKWQTKPKLTIHGDLYYEGKEFETRLKEKKPGDLSDELRISLGMPVGPNAHKVPPPWLIAMQRYGPPPSYPNLKIPGLNSPIPESCSFGYHAGGWGKPPVDETGKPLYGDVFGTNAAEFQTKTEEEEIDRTPWGELEPSDEESSEEEEEEESDEDKPDETGFITPADSGLITPGGFSSVPAGMETPELIELRKKKIEEAMDGSETPQLFTVLPEKRTATVGGAMMGSTHIYDMSTVMSRKGPAPELQGVEVALAPEELELDPMAMTQKYEEHVREQQAQVEKEDFSDMVAEHAAKQKQKKRKAQPQDSRGGSKKYKEFKF

Specifications

Accession Number AAH07610
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10992
Molecular Weight (g/mol) 95.48kDa
Name SF3B2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MDDPSVGPKIPQALEKILQLKESRQEEMNSQQEEEEMETDARSSLGQSASETEEDTVSVSKKEKNRKRRNRKKKKKPQRVRGVSSESSGDREKDSTRSRGSDSPAADVEIEYVTEEPEIYEPNFIFFKRIFEAFKLTDDVKKEKEKEPEKLDKLENSAAPKKKGFEEEHKDSDDDSSDDEQEKKPEAPKLSKKKLRRMNRFTVAELKQLVARPDVVEMHDVTAQDPKLLVHLKATRNSVPVPRHWCFKRKYLQGKRGIEKPPFELPDFIKRTGIQEMREALQEKEEQKTMKSKMREKVRPKMGKIDIDYQKLHDAFFKWQTKPKLTIHGDLYYEGKEFETRLKEKKPGDLSDELRISLGMPVGPNAHKVPPPWLIAMQRYGPPPSYPNLKIPGLNSPIPESCSFGYHAGGWGKPPVDETGKPLYGDVFGTNAAEFQTKTEEEEIDRTPWGELEPSDEESSEEEEEEESDEDKPDETGFITPADSGLITPGGFSSVPAGMETPELIELRKKKIEEAMDGSETPQLFTVLPEKRTATVGGAMMGSTHIYDMSTVMSRKGPAPELQGVEVALAPEELELDPMAMTQKYEEHVREQQAQVEKEDFSDMVAEHAAKQKQKKRKAQPQDSRGGSKKYKEFKF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias SAP145/SF3B145/SF3b1/SF3b150
Common Name SF3B2
Gene Symbol SF3B2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.