Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human sFRP-3 ELISA Kit

Catalog No. EH419RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH419RB 96 Tests
EH419RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH419RB Supplier Invitrogen™ Supplier No. EH419RB
Add to Cart
Add to Cart

ELISA

Human sFRP-3 quantitates human sFRP-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human sFRP-3.

The protein encoded by this gene is a secreted protein that is involved in the regulation of bone development. Defects in this gene are a cause of female-specific osteoarthritis (OA) susceptibility.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Fiz; Fre; FREOS1; frezzled; Fritz; frizzled homolog-related; frizzled-related protein; frizzled-related protein 1; FRP; FRP-3; FRZB; FRZB1; FrzB-1; FRZB1sFRP-3; FRZB-PEN; FZRB; hFIZ; OS1; Secreted frizzled-related protein 3; secreted frizzled-related sequence protein 3; Sfrp3; sFRP-3; SFRP3frezzled; SRFP3
Assay Range 190 to 12,000 pg/mL
Assay Sensitivity 190 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2487
Gene Symbol FRZB
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target FRZB
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.