Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SFRS6 Full-length ORF (NP_006266.2, 1 a.a. - 344 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961672 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-672 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-961-672 Supplier Abnova™ Supplier No. H00006431P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is involved in mRNA splicing and may play a role in the determination of alternative splicing. The encoded nuclear protein belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12. [provided by RefSeq]

Sequence: MPRVYIGRLSYNVREKDIQRFFSGYGRLLEVDLKNGYGFVEFEDSRDADDAVYELNGKELCGERVIVEHARGPRRDRDGYSYGSRSGGGGYSSRRTSGRDKYGPPVRTEYRLIVENLSSRCSWQDLKDFMRQAGEVTYADAHKERTNEGVIEFRSYSDMKRALDKLDGTEINGRNIRLIEDKPRTSHRRSYSGSRSRSRSRRRSRSRSRRSSRSRSRSISKSRSRSRSRSKGRSRSRSKGRKSRSKSKSKPKSDRGSHSHSRSRSKDEYEKSRSRSRSRSPKENGKGDIKSKSRSRSQSRSNSPLPVPPSKARSVSPPPKRATSRSRSRSRSKSRSRSRSSSRD

Specifications

Accession Number NP_006266.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6431
Molecular Weight (g/mol) 66kDa
Name SFRS6 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MPRVYIGRLSYNVREKDIQRFFSGYGRLLEVDLKNGYGFVEFEDSRDADDAVYELNGKELCGERVIVEHARGPRRDRDGYSYGSRSGGGGYSSRRTSGRDKYGPPVRTEYRLIVENLSSRCSWQDLKDFMRQAGEVTYADAHKERTNEGVIEFRSYSDMKRALDKLDGTEINGRNIRLIEDKPRTSHRRSYSGSRSRSRSRRRSRSRSRRSSRSRSRSISKSRSRSRSRSKGRSRSRSKGRKSRSKSKSKPKSDRGSHSHSRSRSKDEYEKSRSRSRSRSPKENGKGDIKSKSRSRSQSRSNSPLPVPPSKARSVSPPPKRATSRSRSRSRSKSRSRSRSSSRD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias B52/FLJ08061/MGC5045/SRP55
Common Name SFRS6
Gene Symbol SFRS6
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.