Learn More
Description
- SFTPA1 surfactant protein A1 [ Homo sapiens (human) ]
- Also known as: SPA; PSAP; PSPA; SP-A; SPA1; PSP-A; SFTP1; SP-A1; COLEC4; SFTPA1B
- The Surfactant Protein A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- SP-A1 is a lung surfactant protein that is a member of a subfamily of C-type lectins called collectins.
- It binds specific carbohydrate moieties found on lipids and on the surface of microorganisms.
- SP-A1 plays an essential role in surfactant homeostasis and in the defense against respiratory pathogens.
- Mutations in SP-A1 are associated with idiopathic pulmonary fibrosis.
- Alternatively spliced transcripts encoding different proteins have been described.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hSFTPA1 |
| Accession Number | NP_005402 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 760 bp |
| Gene ID (Entrez) | 653509 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
