Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SGCA Partial ORF (NP_000014.1, 26 a.a. - 133 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961707 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-707 10 ug
89-961-708 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-707 Supplier Abnova™ Supplier No. H00006442Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a component of the dystrophin-glycoprotein complex (DGC), which is critical to the stability of muscle fiber membranes and to the linking of the actin cytoskeleton to the extracellular matrix. Its expression is thought to be restricted to striated muscle. Mutations in this gene result in type 2D autosomal recessive limb-girdle muscular dystrophy. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: TTLHPLVGRVFVHTLDHETFLSLPEHVAVPPAVHITYHAHLQGHPDLPRWLRYTQRSPHHPGFLYGSATPEDRGLQVIEVTAYNRDSFDTTRQRLVLEIGDPEGPLLP

Specifications

Accession Number NP_000014.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6442
Molecular Weight (g/mol) 37.62kDa
Name SGCA (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen TTLHPLVGRVFVHTLDHETFLSLPEHVAVPPAVHITYHAHLQGHPDLPRWLRYTQRSPHHPGFLYGSATPEDRGLQVIEVTAYNRDSFDTTRQRLVLEIGDPEGPLLP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias 50-DAG/A2/ADL/DAG2/DMDA2/LGMD2D/SCARMD1/adhalin
Common Name SGCA
Gene Symbol SGCA
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.