Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SGCG Partial ORF (NP_000222.1, 191 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961716 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-716 10 ug
89-961-717 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-716 Supplier Abnova™ Supplier No. H00006445Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes gamma-sarcoglycan, one of several sarcolemmal transmembrane glycoproteins that interact with dystrophin. The dystrophin-glycoprotein complex (DGC) spans the sarcolemma and is comprised of dystrophin, syntrophin, alpha- and beta-dystroglycans and sarcoglycans. The DGC provides a structural link between the subsarcolemmal cytoskeleton and the extracellular matrix of muscle cells. Defects in the encoded protein can lead to early onset autosomal recessive muscular dystrophy, in particular limb-girdle muscular dystrophy, type 2C (LGMD2C). [provided by RefSeq]

Sequence: DLRLESPTRSLSMDAPRGVHIQAHAGKIEALSQMDILFHSSDGMLVLDAETVCLPKLVQGTWGPSGSSQSLYEICVCPDGKLYLSVAGVSTTCQEHSHIC

Specifications

Accession Number NP_000222.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6445
Molecular Weight (g/mol) 36.74kDa
Name SGCG (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen DLRLESPTRSLSMDAPRGVHIQAHAGKIEALSQMDILFHSSDGMLVLDAETVCLPKLVQGTWGPSGSSQSLYEICVCPDGKLYLSVAGVSTTCQEHSHIC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias A4/DAGA4/DMDA/DMDA1/LGMD2C/MAM/MGC130048/SCARMD2/SCG3/TYPE
Common Name SGCG
Gene Symbol SGCG
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.