Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SGKL Full-length ORF (AAH15326, 1 a.a. - 496 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961745 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-745 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-961-745 Supplier Abnova™ Supplier No. H00023678P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, KA, WB-Re

This gene is a member of the Ser/Thr protein kinase family and encodes a phosphoprotein with a PX (phox homology) domain. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: MQRDHTMDYKESCPSVSIPSSDEHREKKKRFTVYKVLVSVGRSEWFVFRRYAEFDKLYNTLKKQFPAMALKIPAKRIFGDNFDPDFIKQRRAGLNEFIQNLVRYPELYNHPDVRAFLQMDSPKHQSDPSEDEDERSSQKLHSTSQNINLGPSGNPHAKPTDFDFLKVIGKGSFGKVLLAKRKLDGKFYAVKVLQKKIVLNRKEQKHIMAERNVLLKNVKHPFLVGLHYSFQTTEKLYFVLDFVNGGELFFHLQRERSFPEHRARFYAAEIASALGYLHSIKIVYRDLKPENILLDSVGHVVLTDFGLCKEGIAISDTTTTFCGTPEYLAPEVIRKQPYDNTVDWWCLGAVLYEMLYGLPPFYCRDVAEMYDNILHKPLSLRPGVSLTAWSILEELLEKDRQNRLGAKEDFLEIQNHPFFESLSWADLVQKKIPPPFNPNVAGPDDIRNFDTAFTEETVPYSVCVSSDYSIVNASVLEADDAFVGFSYAPPSEDLFL

Specifications

Accession Number AAH15326
For Use With (Application) Antibody Production, Protein Array, ELISA, Kinase Assay, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23678
Molecular Weight (g/mol) 80.3kDa
Name SGKL (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MQRDHTMDYKESCPSVSIPSSDEHREKKKRFTVYKVLVSVGRSEWFVFRRYAEFDKLYNTLKKQFPAMALKIPAKRIFGDNFDPDFIKQRRAGLNEFIQNLVRYPELYNHPDVRAFLQMDSPKHQSDPSEDEDERSSQKLHSTSQNINLGPSGNPHAKPTDFDFLKVIGKGSFGKVLLAKRKLDGKFYAVKVLQKKIVLNRKEQKHIMAERNVLLKNVKHPFLVGLHYSFQTTEKLYFVLDFVNGGELFFHLQRERSFPEHRARFYAAEIASALGYLHSIKIVYRDLKPENILLDSVGHVVLTDFGLCKEGIAISDTTTTFCGTPEYLAPEVIRKQPYDNTVDWWCLGAVLYEMLYGLPPFYCRDVAEMYDNILHKPLSLRPGVSLTAWSILEELLEKDRQNRLGAKEDFLEIQNHPFFESLSWADLVQKKIPPPFNPNVAGPDDIRNFDTAFTEETVPYSVCVSSDYSIVNASVLEADDAFVGFSYAPPSEDLFL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CISK/DKFZp781N0293/SGK2/SGKL
Common Name SGK3
Gene Symbol SGK3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.