Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SGSM1 Synthetic Peptide

Catalog No. PEP1480
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1480 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1480 Supplier Invitrogen™ Supplier No. PEP1480
Add to Cart
Add to Cart

Synthetic Peptide

SGSM1 Synthetic Peptide, PEP-1480, from Invitrogen.

Small G proteins such as RAP and RAB proteins are the key molecules in intracellular signal transduction and vesicle transportation. A novel protein family small G protein signaling modulator (SGSM) consisting of three members SGSM1-3 bind to RAP and RAB family proteins. All three SGSM proteins possess both a RUN domain and a TBC domain. SGSM1 (RUTBC2) is a 1, 148 amino acid protein that localizes to the Golgi apparatus and is mainly expressed in the CNS. SGSM1 interacts with RAP and RAB subfamily members of the small G proteins, and function as modulators of RAP and RAB-mediated neuronal signal transduction and vesicular transportation pathways.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 129049
Species Human
Purification Method purified
Accession Number Q2NKQ1
Common Name Human SGSM1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol SGSM1
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.