Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SH3BP2 Partial ORF (AAH22996, 452 a.a. - 561 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961792 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-792 25 ug
89-961-791 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-792 Supplier Abnova™ Supplier No. H00006452Q01L

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene has an N-terminal pleckstrin homology (PH) domain, an SH3-binding proline-rich region, and a C-terminal SH2 domain. The protein binds to the SH3 domains of several proteins including the ABL1 and SYK protein tyrosine kinases , and functions as a cytoplasmic adaptor protein to positively regulate transcriptional activity in T, natural killer (NK), and basophilic cells. Mutations in this gene result in cherubism. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: PLPNSVFVNTTESCEVERLFKATSPRGEPQDGLYCIRNSSTKSGKVLVVWDETSNKVRNYRIFEKDSKFYLEGEVLFVSVGSMVEHYHTHVLPSHQSLLLRHPYGYTGPR

Specifications

Accession Number AAH22996
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6452
Molecular Weight (g/mol) 37.73kDa
Name SH3BP2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen PLPNSVFVNTTESCEVERLFKATSPRGEPQDGLYCIRNSSTKSGKVLVVWDETSNKVRNYRIFEKDSKFYLEGEVLFVSVGSMVEHYHTHVLPSHQSLLLRHPYGYTGPR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias 3BP2/CRBM/CRPM/FLJ42079/RES4-23
Common Name SH3BP2
Gene Symbol SH3BP2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.