Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SH3KBP1 Full-length ORF (AAH15806.1, 1 a.a. - 665 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961823 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-823 25 μg
89-961-822 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-823 Supplier Abnova™ Supplier No. H00030011P01L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for AP, Array, ELISA, WB-Re

CBL (MIM 165360) constitutively interacts with SH3 domain-containing proteins and, upon tyrosine phosphorylation, with SH2 domain-containing proteins. The SH3KBP1 gene encodes an 85-kD CBL-interacting protein that enhances tumor necrosis factor (MIM 191160)-mediated apoptotic cell death (Narita et al., 2001 [PubMed 11474197]).[supplied by OMIM]

Sequence: MVEAIVEFDYQAQHDDELTISVGEIITNIRKEDGGWWEGQINGRRGLFPDNFVREIKKEMKKDPLTNKAPEKPLHEVPSGNSLLSSETILRTNKRGERRRRRCQVAFSYLPQNDDELELKVGDIIEVVGEVEEGWWEGVLNGKTGMFPSNFIKELSGESDELGISQDEQLSKSSLRETTGSESDGGDSSSTKSEGANGTVATAAIQPKKVKGVGFGDIFKDKPIKLRPRSIEVENDFLPVEKTIGKKLPATTATPDSSKTEMDSRTKSKDYCKVIFPYEAQNDDELTIKEGDIVTLINKDCIDVGWWEGELNGRRGVFPDNFVKLLPPDFEKEGNRPKKPPPPSAPVIKQGAGTTERKHEIKKIPPERPEMLPNRTEEKERPEREPKLDLQKPSVPAIPPKKPRPPKTNSLSRPGALPPRRPERPVGPLTHTRGDSPKIDLAGSSLSGILDKDLSDRSNDIDLEGFDSVVSSTEKLSHPTTSRPKATGRRPPSQSLTSSSLSSPDIFDSPSPEEDKEEHISLAHRGVDASKKTSKTVTISQVSDNKASLPPKPGTMAAGGGGPAPLSSAVPSPLSSSLGTAGHRANSPSLFGTEGKPKMEPAASSQAAVEELRTQVRELRSIIETMKDQQKREIKQLLSELDEEKKIRLRLQMEVNDIKKALQSK

Specifications

Accession Number AAH15806.1
For Use With (Application) Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 30011
Molecular Weight (g/mol) 99.6kDa
Name Human SH3KBP1 Full-length ORF (AAH15806.1, 1 a.a. - 665 a.a.) Recombinant Protein with GST-tag at N-terminal
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Storage Requirements Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CIN85, GIG10, MIG18
Common Name SH3KBP1
Gene Symbol SH3KBP1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST Tag
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.