Learn More
ACROBiosystems ACROBiosystems Human Siglec-10 Protein, Mouse IgG2a Fc Tag, low endotoxin
Supplier:  ACROBiosystems SI0H525B100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Siglec-10, Mouse IgG2a Fc Tag, low endotoxin (SI0-H525b) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession # Q96LC7-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 85.6 kDa. The protein migrates as 90-120 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-10, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. SI0-H525b) at 5 μg/mL (100 μL/well) can bind Anti-Human Siglec-10 MAb, human IgG with a linear range of 0.6-20 ng/mL (QC tested).
Catalog No. 50-210-6671
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.