Learn More
ACROBiosystems Human Siglec-2 10mg
Supplier:  ACROBiosystems MBSC00710MG
Encompass_Preferred
ActiveMax Human CD22 uBeads premium grade (for cells) are uniform superparamagnetic beads of 5.5 µm with streptavidin on its surface and coupled with biotinylated Human CD22 Protein expressed from human 293 cells (HEK293) and contains AA Asp 20 - Arg 687 (Accession # P20273-1).
Catalog No. 50-253-4465
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.