Learn More
ACROBiosystems ACROBiosystems Human Siglec-2 / CD22 Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems CD2H5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Siglec-2, Fc Tag (CD2-H5253) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 687 (Accession # P20273-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 101.6 kDa. The protein migrates as 125-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-2, Fc Tag (Cat. No. CD2-H5253) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human Siglec-2 Antibody, Human IgG1 with a linear range of 1-20 ng/mL (QC tested).
Catalog No. 50-210-7293
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.