Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Siglec-2 (CD22) (soluble) Recombinant Protein, PeproTech®

Catalog No. 100015UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
100-01-5UG 5 μg
10001100UG 100 μg
100-01-1MG 1 mg
100-01-20UG 20 μg
10001250UG 250 μg
10001500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 100-01-5UG Supplier Gibco™ Supplier No. 100015UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human CD22 is a soluble 75.0 kDa protein which corresponds to the extracellular domain of CD22. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CD22 (BL-CAM) is a type 1 integral membrane glycoprotein with molecular weight of 130 to 140 kDa. CD22 is expressed in both the cytoplasm and cell membrane of B-lymphocytes. CD22 antigen appears early in B-cell lymphocyte differentiation at approximately the same stage as the CD19 antigen. Unlike other B-cell markers, CD22 membrane expression is limited to the late differentiation stages comprised between mature B cells (CD22+) and plasma cells (CD22-), and may thus prove useful in phenotyping mature leukemias. CD22 is also strongly expressed in hairy cell leukemia. CD22 preferentially binds to alpha2,6-linked sialic acid. The sialic acid recognition site can be masked by cis interactions with sialic acids on the same cell surface. Upon ligand induced tyrosine phosphorylation in the immune response seems to be involved in regulation of B cell antigen receptor signaling. CD22 plays a role in positive regulation through interaction with Src family tyrosine kinases and may also act as an inhibitory receptor by recruiting cytoplasmic phosphatases via their SH2 domains that block signal transduction through dephosphorylation of signaling molecules. CD22 is also strongly expressed in hairy cell leukemia. CD22 is also positive in diffuse large B-cell lymphoma and nodular lymphocyte predominance Hodgkin’s lymphoma, but negative in classical Hodgkin’s lymphoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20273
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 933
Molecular Weight (g/mol) 75 kDa
Name Human Siglec-2 (CD22) (soluble)
Quantity 5 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14
Common Name CD22
Gene Symbol CD22
Biological Activity Determined by its ability to inhibit the proliferation of Raji cells. The expected ED50 for this effect is 10-17 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence SKWVFEHPET LYAWEGACVW IPCTYRALDG DLESFILFHN PEYNKNTSKF DGTRLYESTK DGKVPSEQKR VQFLGDKNKN CTLSIHPVHL NDSGQLGLRM ESKTEKWMER IHLNVSERPF PPHIQLPPEI QESQEVTLTC LLNFSCYGYP IQLQWLLEGV PMRQAAVTST SLTIKSVFTR SELKFSPQWS HHGKIVTCQL QDADGKFLSN DTVQLNVKHT PKLEIKVTPS DAIVREGDSV TMTCEVSSSN PEYTTVSWLK DGTSLKKQNT FTLNLREVTK DQSGKYCCQV SNDVGPGRSE EVFLQVQYAP EPSTVQILHS PAVEGSQVEF LCMSLANPLP TNYTWYHNGK EMQGRTEEKV HIPKILPWHA GTYSCVAENI LGTGQRGPGA ELDVQYPPKK VTTVIQNPMP IREGDTVTLS CNYNSSNPSV TRYEWKPHGA WEEPSLGVLK IQNVGWDNTT IACARCNSWC SWASPVALNV QYAPRDVRVR KIKPLSEIHS GNSVSLQCDF SSSHPKEVQF FWEKNGRLLG KESQLNFDSI SPEDAGSYSC WVNNSIGQTA SKAWTLEVLY APRRLRVSMS PGDQVMEGKS ATLTCESDAN PPVSHYTWFD WNNQSLPHHS QKLRLEPVKV QHSGAYWCQG TNSVGKGRSP LSTLTVYYSP ETIGRR
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.