Learn More
ACROBiosystems Human Siglec-3 100ug
Supplier:  ACROBiosystems CD3H5258100UG
Encompass_Preferred
Human Siglec-3 (140-232) Llama IgG2b Fc Tag (CD3-H5258) is expressed from human 293 cells (HEK293). It contains AA Asp 140 - Thr 232 (Accession # P20138-1).
Catalog No. 50-253-5384
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.