Learn More
Bon Opus Biosciences Recombinant Human Siglec-5/CD170 (C-His) - Source: Human 293 Cells
SDP
Supplier:  Bon Opus Biosciences BP26850UG
Encompass_Preferred
Recombinant Human Sialic Acid-binding Ig-like Lectin 5 is produced by our Mammalian expression system and the target gene encoding Glu17-Thr434 is expressed with a 6His at the C-terminus. Accession#O15389. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4
Catalog No. NC2733753
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.