Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Siglec-9 Fluorescein-conjugated Antibody, R&D Systems™

Catalog No. p-4717686
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1139F025 25 Tests
FAB1139F100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1139F025 Supplier R&D Systems Supplier No. FAB1139F025
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-9
Applications Flow Cytometry
Classification Monoclonal
Clone 191240
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9Y336
Gene Alias CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9
Gene Symbols SIGLEC9
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-9 Gln18-Gly348 (predicted) Accession # Q9Y336
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27180
Test Specificity Detects human Siglec-9 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -5, -7 or -10 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.