Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SIGLEC12 Full-length ORF (NP_443729.1, 1 a.a. - 595 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961911 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-911 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-961-911 Supplier Abnova™ Supplier No. H00089858P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Sialic acid-binding immunoglobulin-like lectins (SIGLECs) are a family of cell surface proteins belonging to the immunoglobulin superfamily. They mediate protein-carbohydrate interactions by selectively binding to different sialic acid moieties present on glycolipids and glycoproteins. This gene encodes a member of the SIGLEC3-like subfamily of SIGLECs. Members of this subfamily are characterized by an extracellular V-set immunoglobulin-like domain followed by two C2-set immunoglobulin-like domains, and the cytoplasmic tyrosine-based motifs ITIM and SLAM-like. The encoded protein, upon tyrosine phosphorylation, has been shown to recruit the Src homology 2 domain-containing protein-tyrosine phosphatases SHP1 and SHP2. It has been suggested that the protein is involved in the negative regulation of macrophage signaling by functioning as an inhibitory receptor. This gene is located in a cluster with other SIGLEC3-like genes on 19q13.4. Alternatively spliced transcript variants encoding distinct isoforms have been described for this gene. [provided by RefSeq]

Sequence: MLLLLLLLPPLLCGRVGAKEQKDYLLTMQKSVTVQEGLCVSVLCSFSYPQNGWTASDPVHGYWFRAGDHVSRNIPVATNNPARAVQEETRDRFHLLGDPQNKDCTLSIRDTRESDAGTYVFCVERGNMKWNYKYDQLSVNVTASQDLLSRYRLEVPESVTVQEGLCVSVPCSVLYPHYNWTASSPVYGSWFKEGADIPWDIPVATNTPSGKVQEDTHGRFLLLGDPQTNNCSLSIRDARKGDSGKYYFQVERGSRKWNYIYDKLSVHVTALTHMPTFSIPGTLESGHPRNLTCSVPWACEQGTPPTITWMGASVSSLDPTITRSSMLSLIPQPQDHGTSLTCQVTLPGAGVTMTRAVRLNISYPPQNLTMTVFQGDGTASTTLRNGSALSVLEGQSLHLVCAVDSNPPARLSWTWGSLTLSPSQSSNLGVLELPRVHVKDEGEFTCRAQNPLGSQHISLSLSLQNEYTGKMRPISGVTLGAFGGAGATALVFLYFCIIFVVVRSCRKKSARPAVGVGDTGMEDANAVRGSASQGPLIESPADDSPPHHAPPALATPSPEEGEIQYASLSFHKARPQYPQEQEAIGYEYSEINIPK

Specifications

Accession Number NP_443729.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 89858
Molecular Weight (g/mol) 91.4kDa
Name SIGLEC12 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MLLLLLLLPPLLCGRVGAKEQKDYLLTMQKSVTVQEGLCVSVLCSFSYPQNGWTASDPVHGYWFRAGDHVSRNIPVATNNPARAVQEETRDRFHLLGDPQNKDCTLSIRDTRESDAGTYVFCVERGNMKWNYKYDQLSVNVTASQDLLSRYRLEVPESVTVQEGLCVSVPCSVLYPHYNWTASSPVYGSWFKEGADIPWDIPVATNTPSGKVQEDTHGRFLLLGDPQTNNCSLSIRDARKGDSGKYYFQVERGSRKWNYIYDKLSVHVTALTHMPTFSIPGTLESGHPRNLTCSVPWACEQGTPPTITWMGASVSSLDPTITRSSMLSLIPQPQDHGTSLTCQVTLPGAGVTMTRAVRLNISYPPQNLTMTVFQGDGTASTTLRNGSALSVLEGQSLHLVCAVDSNPPARLSWTWGSLTLSPSQSSNLGVLELPRVHVKDEGEFTCRAQNPLGSQHISLSLSLQNEYTGKMRPISGVTLGAFGGAGATALVFLYFCIIFVVVRSCRKKSARPAVGVGDTGMEDANAVRGSASQGPLIESPADDSPPHHAPPALATPSPEEGEIQYASLSFHKARPQYPQEQEAIGYEYSEINIPK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ38600/S2V/SIGLECL1/SLG/Siglec-12/Siglec-L1/Siglec-XII
Common Name SIGLEC12
Gene Symbol SIGLEC12
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.