Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SIM1 Partial ORF (NP_005059, 657 a.a. - 766 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961942 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-942 10 ug
89-961-943 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-942 Supplier Abnova™ Supplier No. H00006492Q01S

Used for AP, Array, ELISA, WB-Re

SIM1 and SIM2 genes are Drosophila single-minded (sim) gene homologs. SIM1 transcript was detected only in fetal kidney out of various adult and fetal tissues tested. Since the sim gene plays an important role in Drosophila development and has peak levels of expression during the period of neurogenesis,it was proposed that the human SIM gene is a candidate for involvement in certain dysmorphic features (particularly the facial and skull characteristics), abnormalities of brain development, and/or mental retardation of Down syndrome. [provided by RefSeq]

Sequence: RISSPNSDRISKSSLILAKDYLHSDISPHQTAGDHPTVSPNCFGSHRQYFDKHAYTLTGYALEHLYDSETIRNYSLGCNGSHFDVTSHLRMQPDPAQGHKGTSVIITNGS

Specifications

Accession Number NP_005059
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6492
Molecular Weight (g/mol) 37.84kDa
Name SIM1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen RISSPNSDRISKSSLILAKDYLHSDISPHQTAGDHPTVSPNCFGSHRQYFDKHAYTLTGYALEHLYDSETIRNYSLGCNGSHFDVTSHLRMQPDPAQGHKGTSVIITNGS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias bHLHe14
Common Name SIM1
Gene Symbol SIM1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.