Learn More
ACROBiosystems ACROBiosystems Human SIRP alphaV8 / CD172a Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems SI8H52H5100UG
Encompass_Preferred
Recombinant Protein;100UG;Human SIRP alphaV8, His Tag (SI8-H52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 369 (Accession # ATD50864.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.1 kDa. The protein migrates as 46-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alphaV8, His Tag (Cat. No. SI8-H52H5) at 2 μg/mL (100 μL/well) can bind Human CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-H52A5) with a linear range of 1-5 ng/mL (QC tested).
Catalog No. 50-210-6458
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.