Learn More
ACROBiosystems Biotinylated Human SIRP gamma, His,Avitag (SIG-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 29 - Ser 364 (Accession # AAH64532.1).
Supplier:  ACROBiosystems SIGH82E325UG
Encompass_Preferred
Signal-regulatory protein gamma (SIRPG or SIRP-gamma) is also known as CD172 antigen-like family member B (CD172g), Signal-regulatory protein beta-2 (SIRPB2), is a single-pass type I membrane protein. SIRPG is expressed on CD4+ T-cells, CD8+ T-cells, CD56-bright natural killer (NK) cells, CD20+ cells, and all activated NK cells. SIRPG interacts with CD47. On binding with CD47, CD172g mediates cell-cell adhesion.
Catalog No. 50-223-1325
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.