Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SIT1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5016SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5016SP 25 μg
AF5016 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF5016SP Supplier R&D Systems Supplier No. AF5016SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SIT1 Polyclonal specifically detects SIT1 in Human samples. It is validated for Western Blot.

Specifications

Antigen SIT1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y3P8
Gene Alias gp30/40, MGC125908, MGC125909, RP11-331F9.5, SHP2 interacting transmembrane adaptor, SHP2-interacting transmembrane adapter protein, SHP2-interacting transmembrane adaptor protein, signaling threshold regulating transmembrane adaptor 1, signaling threshold-regulating transmembrane adapter 1, SITMGC125910, suppression inducing transmembrane adaptor 1, Suppression-inducing transmembrane adapter 1
Gene Symbols SIT1
Host Species Goat
Immunogen E. coli-derived recombinant human SIT1 Gln65-Ser196 Accession # Q9Y3P8
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27240
Test Specificity Detects human SIT1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.