Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Human Skull Model with Muscles
SCI_ED

Catalog No. S81007 Shop All Walter Products Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S81007 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81007 Supplier Walter Products™ Supplier No. B10208
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 3 part model has numerical markings for easy identification of the cranial bones, sutures and muscles

  • Medical quality anatomical model
  • Skull separates into 3 parts: the skull cap and mandible are removable for further study
  • Skull cap is magnetic and can be removed for further study of the inner cranial bones
  • Jaw is spring mounted to show natural movement of the mandible
  • Features hand painted sutures in orange, muscle origins in red and insertions in blue
  • Identification key card included

Specifications

For Use With (Application) Education
Height (Metric) 15 cm
Includes Skull & manual
Length (Metric) 15 cm
Material PVC
Quantity 1
Size Life-size
Weight (Metric) 1
Width (Metric) 23 cm
Product Type Model, Human Skull
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.