Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC11A1 Full-length ORF (AAH41787.1, 1 a.a. - 178 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962043 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-043 25 ug
89-962-042 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-043 Supplier Abnova™ Supplier No. H00006556P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the solute carrier family 11 (proton-coupled divalent metal ion transporters) family and encodes a multi-pass membrane protein. The protein functions as a divalent transition metal (iron and manganese) transporter involved in iron metabolism and host resistance to certain pathogens. Mutations in this gene have been associated with susceptibility to infectious diseases such as tuberculosis and leprosy, and inflammatory diseases such as rheumatoid arthritis and Crohn disease. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined. [provided by RefSeq]

Sequence: MTGDKGPQRLSGSSYGSISSPTSPTSPGPQQAPPRETYLRNIESDLQAGAVAGFKLLWVLLWATVLGLLCQRLAARLGVVTGKDLGEVCHLYYPKVPRTVLWLTIELAIVGSDMQEVIGTAIAFNLLSAGRYHPSVPQLFRPGREQLLLLPPLTSPSQSLFYPAVPSEAGLLPCFPEM

Specifications

Accession Number AAH41787.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6556
Molecular Weight (g/mol) 45.6kDa
Name SLC11A1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MTGDKGPQRLSGSSYGSISSPTSPTSPGPQQAPPRETYLRNIESDLQAGAVAGFKLLWVLLWATVLGLLCQRLAARLGVVTGKDLGEVCHLYYPKVPRTVLWLTIELAIVGSDMQEVIGTAIAFNLLSAGRYHPSVPQLFRPGREQLLLLPPLTSPSQSLFYPAVPSEAGLLPCFPEM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias LSH/NRAMP/NRAMP1
Common Name SLC11A1
Gene Symbol SLC11A1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.