Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC11A2 Partial ORF (NP_000608, 1 a.a. - 65 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962044 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-044 10 ug
89-962-045 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-044 Supplier Abnova™ Supplier No. H00004891Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The SLC11A2 gene encodes a divalent metal transporter (DMT1), which carries iron, manganese, cobalt, nickel, cadmium, lead, copper, and zinc. DMT1 participates in cellular iron absorption at the luminal surface of the duodenum as well as in other areas of the body (Hubert and Hentze, 2002 [PubMed 12209011]; Ludwiczek et al., 2007 [PubMed 17293870]).[supplied by OMIM]

Sequence: MVLGPEQKMSDDSVSGDHGESASLGNINPAYSNPSLSQSPGDSEEYFATYFNEKISIPEEEYSCF

Specifications

Accession Number NP_000608
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4891
Molecular Weight (g/mol) 32.89kDa
Name SLC11A2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MVLGPEQKMSDDSVSGDHGESASLGNINPAYSNPSLSQSPGDSEEYFATYFNEKISIPEEEYSCF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DCT1/DMT1/FLJ37416/NRAMP2
Common Name SLC11A2
Gene Symbol SLC11A2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.