Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC12A1 Full-length ORF (ENSP00000331550, 1 a.a. - 430 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962048 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-048 10 ug
89-962-049 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-048 Supplier Abnova™ Supplier No. H00006557P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The sodium-potassium-chloride cotransporter isoform 2 is kidney-specific and is found on the apical membrane of the thick ascending limb of Henle's loop and the macula densa. It accounts for most of the NaCl resorption with the stoichiometry of 1Na:1K:2Cl and is sensitive to such diuretics as furosemide and bumetanide. Some Bartter-like syndromes result from defects in this gene. [provided by RefSeq]

Sequence: MSLNNSSNVFLDSVPSNTNRFQVSVINENHESSAAADDNTDPPHYEETSFGDEAQKRLRISFRPGNQECYDNFLQSGETAKTDASFHAYDSHTNTYYLQTFGHNTMDAVPKIEYYRNTGSISGPKVNRPSLLEIHEQLAKNVAVTPSSADRVANGDGIPGDEQAENKEDDQAGVVKFGWVKGVLVRCMLNIWGVMLFIRLSWIVGEAGIGLGVLIILLSTMVTSITGLSTSAIATNGFVRGGGAYYLISRSLGPEFGGSIGLIFAFANAVAVAMYVVGFAETVVDLLKESDSMMVDPTNDIRIIGSITVVILLGISVAGMEWEAKAQVILLVILLIAIANFFIGTVIPSNNEKKSRGFFNYQASIFAENFGPRFTKGEGFFSVFAIFFPAATGILAGANISGDLEALRKQGASPLPQGAQSRRRRQTSLI

Specifications

Accession Number ENSP00000331550
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6557
Molecular Weight (g/mol) 72.9kDa
Name SLC12A1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MSLNNSSNVFLDSVPSNTNRFQVSVINENHESSAAADDNTDPPHYEETSFGDEAQKRLRISFRPGNQECYDNFLQSGETAKTDASFHAYDSHTNTYYLQTFGHNTMDAVPKIEYYRNTGSISGPKVNRPSLLEIHEQLAKNVAVTPSSADRVANGDGIPGDEQAENKEDDQAGVVKFGWVKGVLVRCMLNIWGVMLFIRLSWIVGEAGIGLGVLIILLSTMVTSITGLSTSAIATNGFVRGGGAYYLISRSLGPEFGGSIGLIFAFANAVAVAMYVVGFAETVVDLLKESDSMMVDPTNDIRIIGSITVVILLGISVAGMEWEAKAQVILLVILLIAIANFFIGTVIPSNNEKKSRGFFNYQASIFAENFGPRFTKGEGFFSVFAIFFPAATGILAGANISGDLEALRKQGASPLPQGAQSRRRRQTSLI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BSC1/MGC48843/NKCC2
Common Name SLC12A1
Gene Symbol SLC12A1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.