Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC12A2 Partial ORF (NP_001037.1, 903 a.a. - 1010 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962051 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-051 25 ug
89-962-050 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-051 Supplier Abnova™ Supplier No. H00006558Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

By moving chloride into epithelial cells, the Na-K-Cl cotransporter SLC12A2 aids transcellular movement of chloride across both secretory and absorptive epithelia (Payne et al., 1995 [PubMed 7629105]). See also SLC12A1 (MIM 600839) and SLC12A3 (MIM 600968).[supplied by OMIM]

Sequence: YINLFHDAFDIQYGVVVIRLKEGLDISHLQGQEELLSSQEKSPGTKDVVVSVEYSKKSDLDTSKPLSEKPITHKVEEEDGKTATQPLLKKESKGPIVPLNVADQKLLE

Specifications

Accession Number NP_001037.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6558
Molecular Weight (g/mol) 37.62kDa
Name SLC12A2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen YINLFHDAFDIQYGVVVIRLKEGLDISHLQGQEELLSSQEKSPGTKDVVVSVEYSKKSDLDTSKPLSEKPITHKVEEEDGKTATQPLLKKESKGPIVPLNVADQKLLE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BSC/BSC2/MGC104233/NKCC1
Common Name SLC12A2
Gene Symbol SLC12A2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.