Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SLC14A1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7067577
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8238SP 25 μg
MAB8238 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8238SP Supplier R&D Systems Supplier No. MAB8238SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC14A1 Monoclonal specifically detects SLC14A1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen SLC14A1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 888418
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13336
Gene Alias FLJ33745, HsT1341, HUT11, JK, JKFLJ41687, RACH1, RACH2, solute carrier family 14 (urea transporter), member 1 (Kidd blood group), Solute carrier family 14 member 1, truncated urea transporter, urea transporter 1, urea transporter JK glycoprotein, Urea transporter, erythrocyte, urea transporter-B1, UT1, UT1UT-B1, UT-B1, UTE, UTEblood group Kidd urea transporter
Gene Symbols SLC14A1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC14A1 Accession # Q13336
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6563
Test Specificity Detects human SLC14A1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.