Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC16A3 Full-length ORF (ADR83188.1) Recombinant Protein without tag

Catalog No. 89962077 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-077 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-077 Supplier Abnova™ Supplier No. H00009123G01

Used for AP, Func, Screening

Lactic acid and pyruvate transport across plasma membranes is catalyzed by members of the proton-linked monocarboxylate transporter (MCT) family, which has been designated solute carrier family-16. Each MCT appears to have slightly different substrate and inhibitor specificities and transport kinetics, which are related to the metabolic requirements of the tissues in which it is found. The MCTs, which include MCT1 (SLC16A1; MIM 600682) and MCT2 (SLC16A7; MIM 603654), are characterized by 12 predicted transmembrane domains (Price et al., 1998 [PubMed 9425115]).[supplied by OMIM]

Sequence: MGGAVVDEGPTGVKAPDGGWGWAVLFGCFVITGFSYAFPKAVSVFFKELIQEFGIGYSDTAWISSILLAMLYGTGPLCSVCVNRFGCRPVMLVGGLFASLGMVAASFCRSIIQVYLTTGVITGLGLALNFQPSLIMLNRYFSKRRPMANGLAAAGSPVFLCALSPLGQLLQDRYGWRGGFLILGGLLLNCCVCAALMRPLVVTAQPGSGPPRPSRRLLDLSVFRDRGFVLYAVAASVMVLGLFVPPVFVVSYAKDLGVPDTKAAFLLTILGFIDIFARPAAGFVAGLGKVRPYSVYLFSFSMFFNGLADLAGSTAGDYGGLVVFCIFFGISYGMVGALQFEVLMAIVGTHKFSSAIGLVLLMEAVAVLVGPPSGGKLLDATHVYMYVFILAGAEVLTSSLILLLGNFFCIRKKPKEPQPEVAAAEEEKLHKPPADSGVDLREVEHFLKAEPEKNGEVVHTPETSV

Specifications

Accession Number ADR83188.1
For Use With (Application) Antibody Production, Compound Screening, Functional Study
Formulation Liquid
Gene ID (Entrez) 9123
Molecular Weight (g/mol) 51.2kDa
Name SLC16A3 (Human) Recombinant Protein
Quantity 2 μg
Immunogen MGGAVVDEGPTGVKAPDGGWGWAVLFGCFVITGFSYAFPKAVSVFFKELIQEFGIGYSDTAWISSILLAMLYGTGPLCSVCVNRFGCRPVMLVGGLFASLGMVAASFCRSIIQVYLTTGVITGLGLALNFQPSLIMLNRYFSKRRPMANGLAAAGSPVFLCALSPLGQLLQDRYGWRGGFLILGGLLLNCCVCAALMRPLVVTAQPGSGPPRPSRRLLDLSVFRDRGFVLYAVAASVMVLGLFVPPVFVVSYAKDLGVPDTKAAFLLTILGFIDIFARPAAGFVAGLGKVRPYSVYLFSFSMFFNGLADLAGSTAGDYGGLVVFCIFFGISYGMVGALQFEVLMAIVGTHKFSSAIGLVLLMEAVAVLVGPPSGGKLLDATHVYMYVFILAGAEVLTSSLILLLGNFFCIRKKPKEPQPEVAAAEEEKLHKPPADSGVDLREVEHFLKAEPEKNGEVVHTPETSV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MCT3/MCT4/MGC138472/MGC138474
Common Name SLC16A3
Gene Symbol SLC16A3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag None
Expression System wheat germ expression system with proprietary liposome technology
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.