Learn More
Specifications
Specifications
| Gene | SLC16A5 |
| Accession Number | NP_004686.1 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1531 bp |
| Gene ID (Entrez) | 9121 |
| Gene Alias | MCT 5, MCT 6, MCT5, MCT6, monocarboxylate transporter 5, monocarboxylate transporter 6, solute carrier family 16 (monocarboxylic acid transporters), member 5, solute carrier family 16 member 5, solute carrier family 16, member 5 (monocarboxylic acid transporter 6) |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
