Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC22A3 Partial ORF (NP_068812, 90 a.a. - 155 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962132 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-132 10 ug
89-962-133 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-132 Supplier Abnova™ Supplier No. H00006581Q01S

Used for AP, Array, ELISA, WB-Re

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. [provided by RefSeq]

Sequence: CQRYLLEAANDSASATSALSCADPLAAFPNRSAPLVPCRGGWRYAQAHSTIVSEFDLVCVNAWMLD

Specifications

Accession Number NP_068812
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6581
Molecular Weight (g/mol) 33kDa
Name SLC22A3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen CQRYLLEAANDSASATSALSCADPLAAFPNRSAPLVPCRGGWRYAQAHSTIVSEFDLVCVNAWMLD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias EMT/EMTH/OCT3
Common Name SLC22A3
Gene Symbol SLC22A3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.