Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC26A5 Full-length ORF (AAI00836.1, 1 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962224 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-224 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-224 Supplier Abnova™ Supplier No. H00375611P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the SLC26A/SulP transporter family. It encodes a protein that is specifically expressed in outer hair cells (OHCs) of the cochlea and is essential in auditory processing. Intracellular anions are thought to act as extrinsic voltage sensors, which bind to this protein and trigger the conformational changes required for rapid length changes in OHCs. Mutations in this gene have been associated with non-syndromic hearing loss. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: MDHAEENEILAATQRYYVERPIFSHPVLQERLHTKDKVPDSIADKLKQAFTCTPKKIRNIIYMFLPITKWLPAYKFKEYVLGDLVSGISTGVLQLPQGLAFAMLAAVPPIFGLYSSFYPVIMYCFLGTSRHISIGPFAVISLMIGGVAVRLVPDDIVIPGGVNATNGTEARDALRVKVAMSVTLLSGIIQFCLGVCRFGFVAIYLTEPLVRGFTTAAAVHVFTSMLKYLFGVKTKRYSGIFSVVYSTVAVLQNVKNLNVCSLGVGLMVFGLLLGGKEFNERFKEKLPAPIPLEFFAVVMGTGISAGFNLKESYNVDVVGTLPLGLLPPANPDTSLFHLVYVDAIAIAIVGFSVTISMAKTLANKHGYQVDGNQELIALGLCNSIGSLFQTFSISCSLSRSLVQEGTGGKTQTIWLTTFVSSLFLGLDYGLITAVIIALLTVIYRTQR

Specifications

Accession Number AAI00836.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 375611
Molecular Weight (g/mol) 75kDa
Name SLC26A5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MDHAEENEILAATQRYYVERPIFSHPVLQERLHTKDKVPDSIADKLKQAFTCTPKKIRNIIYMFLPITKWLPAYKFKEYVLGDLVSGISTGVLQLPQGLAFAMLAAVPPIFGLYSSFYPVIMYCFLGTSRHISIGPFAVISLMIGGVAVRLVPDDIVIPGGVNATNGTEARDALRVKVAMSVTLLSGIIQFCLGVCRFGFVAIYLTEPLVRGFTTAAAVHVFTSMLKYLFGVKTKRYSGIFSVVYSTVAVLQNVKNLNVCSLGVGLMVFGLLLGGKEFNERFKEKLPAPIPLEFFAVVMGTGISAGFNLKESYNVDVVGTLPLGLLPPANPDTSLFHLVYVDAIAIAIVGFSVTISMAKTLANKHGYQVDGNQELIALGLCNSIGSLFQTFSISCSLSRSLVQEGTGGKTQTIWLTTFVSSLFLGLDYGLITAVIIALLTVIYRTQR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DFNB61/MGC118886/MGC118887/MGC118888/MGC118889/PRES
Common Name SLC26A5
Gene Symbol SLC26A5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.