Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC27A2 Full-length ORF (AAH57770.1, 1 a.a. - 567 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962238 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-238 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-238 Supplier Abnova™ Supplier No. H00011001P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is an isozyme of long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme activates long-chain, branched-chain and very-long-chain fatty acids containing 22 or more carbons to their CoA derivatives. It is expressed primarily in liver and kidney, and is present in both endoplasmic reticulum and peroxisomes, but not in mitochondria. Its decreased peroxisomal enzyme activity is in part responsible for the biochemical pathology in X-linked adrenoleukodystrophy. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MLSAIYTVLAGLLFLPLLVNLCCPYFFQDIGYFLKVAAVGRRVRSYGKRRPARTILRAFLEKARQTPHKPFLLFRDETLTYAQVDRRSNQVARALHDHLGLRQGDCVALLMGNEPAYVWLWLGLVKLGCAMACLNYNIRAKSLLHCFQCCGAKVLLVSPELQAAVEEILPSLKKDDVSIYYVSRTSNTDGIDSFLDKVDEVSTEPIPESWRSEVTFSTPALYIYTSGTTGATLALRTKFSASQFWDDCRKYNVTVIQYIGELLRYLCNSPQKPNDRDHKVRLALGNGLRGDVWRQFVKRFGDICIYEFYAATEGNIGFMNYARKVGAVGRVNYLQKKIITYDLIKYDVEKDEPVRDENGYCVRVPKGEVGLLVCKITQLTPFNGYAGAKAQTEKKKLRDVFKKGDLYFNSGDLLMVDHENFIYFHDRVGDTFRWKGENVATTEVADTVGLVDFVQEVNVYGVHVPDHEGRIGMASIKMKENHEFDGKKLFQHIADYLPSYARPRFLRIQDTIEITGTFKHRKMTLVEEGFNPAVIKDALYFLDDTAKMYVPMTEDIYNAISAKTLKL

Specifications

Accession Number AAH57770.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11001
Molecular Weight (g/mol) 91kDa
Name SLC27A2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MLSAIYTVLAGLLFLPLLVNLCCPYFFQDIGYFLKVAAVGRRVRSYGKRRPARTILRAFLEKARQTPHKPFLLFRDETLTYAQVDRRSNQVARALHDHLGLRQGDCVALLMGNEPAYVWLWLGLVKLGCAMACLNYNIRAKSLLHCFQCCGAKVLLVSPELQAAVEEILPSLKKDDVSIYYVSRTSNTDGIDSFLDKVDEVSTEPIPESWRSEVTFSTPALYIYTSGTTGATLALRTKFSASQFWDDCRKYNVTVIQYIGELLRYLCNSPQKPNDRDHKVRLALGNGLRGDVWRQFVKRFGDICIYEFYAATEGNIGFMNYARKVGAVGRVNYLQKKIITYDLIKYDVEKDEPVRDENGYCVRVPKGEVGLLVCKITQLTPFNGYAGAKAQTEKKKLRDVFKKGDLYFNSGDLLMVDHENFIYFHDRVGDTFRWKGENVATTEVADTVGLVDFVQEVNVYGVHVPDHEGRIGMASIKMKENHEFDGKKLFQHIADYLPSYARPRFLRIQDTIEITGTFKHRKMTLVEEGFNPAVIKDALYFLDDTAKMYVPMTEDIYNAISAKTLKL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ACSVL1/FACVL1/FATP2/HsT17226/VLACS/VLCS/hFACVL1
Common Name SLC27A2
Gene Symbol SLC27A2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.