Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC29A4 Partial ORF (NP_694979, 283 a.a. - 345 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962255 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-255 25 μg
89-962-254 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-255 Supplier Abnova™ Supplier No. H00222962Q01L

Used for AP, Array, ELISA, WB-Re

This gene is a member of the SLC29 family and encodes a plasma membrane protein with 11 transmembrane helices. This protein catalyzes the reuptake of monoamines into presynaptic neurons, thus determining the intensity and duration of monoamine neural signaling. It has been shown to transport several compounds, including serotonin, dopamine, and the neurotoxin 1-methyl-4-phenylpyridinium. Alternate transcriptional splice variants which encode the same protein have been characterized. [provided by RefSeq]

Sequence: VHHDVVAGDVHFEHPAPAPAPNESPKDSPAHEVTGSGGAYMRFDVPRPRVQRSWPTFRALLLH

Specifications

Accession Number NP_694979
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 222962
Molecular Weight (g/mol) 32.67kDa
Name SLC29A4 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen VHHDVVAGDVHFEHPAPAPAPNESPKDSPAHEVTGSGGAYMRFDVPRPRVQRSWPTFRALLLH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ENT4/FLJ34923/PMAT
Common Name SLC29A4
Gene Symbol SLC29A4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.