Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC2A10 Partial ORF (NP_110404, 325 a.a. - 382 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962256 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-256 10 μg
89-962-257 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-256 Supplier Abnova™ Supplier No. H00081031Q01S

Used for AP, Array, ELISA, WB-Re

SLC2A10 is a member of the facilitative glucose transporter family, which plays a significant role in maintaining glucose homeostasis.[supplied by OMIM]

Sequence: SGPSCLAVPNATGQTGLPGDSGLLQDSSLPPIPRTNEDQREPILSTAKKTKPHPRSGD

Specifications

Accession Number NP_110404
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 81031
Molecular Weight (g/mol) 32.12kDa
Name SLC2A10 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen SGPSCLAVPNATGQTGLPGDSGLLQDSSLPPIPRTNEDQREPILSTAKKTKPHPRSGD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ATS/GLUT10/MGC126706
Common Name SLC2A10
Gene Symbol SLC2A10
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.