Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC2A9 Partial ORF (NP_001001290, 224 a.a. - 287 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962273 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-273 25 μg
89-962-272 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-273 Supplier Abnova™ Supplier No. H00056606Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the SLC2A facilitative glucose transporter family. Members of this family play a significant role in maintaining glucose homeostasis. The encoded protein may play a role in the development and survival of chondrocytes in cartilage matrices. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: PDSPRYLLLEKHNEARAVKAFQTFLGKADVSQEVEEVLAESRVQRSIRLVSVLELLRAPYVRWQ

Specifications

Accession Number NP_001001290
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 56606
Molecular Weight (g/mol) 32.78kDa
Name SLC2A9 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen PDSPRYLLLEKHNEARAVKAFQTFLGKADVSQEVEEVLAESRVQRSIRLVSVLELLRAPYVRWQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GLUT9/GLUTX/UAQTL2/URATv1
Common Name SLC2A9
Gene Symbol SLC2A9
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.