Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC33A1 Partial ORF (NP_004724, 1 a.a. - 69 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962277 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-277 10 μg
89-962-278 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-277 Supplier Abnova™ Supplier No. H00009197Q01S

Used for AP, Array, ELISA, WB-Re

The encoded protein is required for the formation of O-acetylated (Ac) gangliosides. It is predicted to contain 6 to 10 transmembrane domains, and a leucine zipper motif in transmembrane domain III. Studies indicate that the protein is localized to the cytoplasm. [provided by RefSeq]

Sequence: MSPTISHKDSSRQRRPGNFSHSLDMKSGPLPPGGWDDSHLDSAGREGDREALLGDTGTGDFLKAPQSFR

Specifications

Accession Number NP_004724
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9197
Molecular Weight (g/mol) 33.33kDa
Name SLC33A1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Immunogen MSPTISHKDSSRQRRPGNFSHSLDMKSGPLPPGGWDDSHLDSAGREGDREALLGDTGTGDFLKAPQSFR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ACATN/AT-1/AT1/SPG42
Common Name SLC33A1
Gene Symbol SLC33A1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.