Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC37A4 Full-length ORF (NP_001458.1, 1 a.a. - 429 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962319 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-319 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-319 Supplier Abnova™ Supplier No. H00002542P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene regulates glucose-6-phosphate transport from the cytoplasm to the lumen of the endoplasmic reticulum, in order to maintain glucose homeostasis. It also plays a role in ATP-mediated calcium sequestration in the lumen of the endoplasmic reticulum. Mutations in this gene have been associated with various forms of glycogen storage disease. Alternative splicing in this gene results in multiple transcript variants

Sequence: MAAQGYGYYRTVIFSAMFGGYSLYYFNRKTFSFVMPSLVEEIPLDKDDLGFITSSQSAAYAISKFVSGVLSDQMSARWLFSSGLLLVGLVNIFFAWSSTVPVFAALWFLNGLAQGLGWPPCGKVLRKWFEPSQFGTWWAILSTSMNLAGGLGPILATILAQSYSWRSTLALSGALCVVVSFLCLLLIHNEPADVGLRNLDPMPSEGKKGSLKEESTLQELLLSPYLWVLSTGYLVVFGVKTCCTDWGQFFLIQEKGQSALVGSSYMSALEVGGLVGSIAAGYLSDRAMAKAGLSNYGNPRHGLLLFMMAGMTVSMYLFRVTVTSDSPKLWILVLGAVFGFSSYGPIALFGVIANESAPPNLCGTSHAIVGLMANVGGFLAGLPFSTIAKHYSWSTAFWVAEVICAASTAAFFLLRNIRTKMGRVSKKAE

Specifications

Accession Number NP_001458.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2542
Molecular Weight (g/mol) 72.8kDa
Name SLC37A4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MAAQGYGYYRTVIFSAMFGGYSLYYFNRKTFSFVMPSLVEEIPLDKDDLGFITSSQSAAYAISKFVSGVLSDQMSARWLFSSGLLLVGLVNIFFAWSSTVPVFAALWFLNGLAQGLGWPPCGKVLRKWFEPSQFGTWWAILSTSMNLAGGLGPILATILAQSYSWRSTLALSGALCVVVSFLCLLLIHNEPADVGLRNLDPMPSEGKKGSLKEESTLQELLLSPYLWVLSTGYLVVFGVKTCCTDWGQFFLIQEKGQSALVGSSYMSALEVGGLVGSIAAGYLSDRAMAKAGLSNYGNPRHGLLLFMMAGMTVSMYLFRVTVTSDSPKLWILVLGAVFGFSSYGPIALFGVIANESAPPNLCGTSHAIVGLMANVGGFLAGLPFSTIAKHYSWSTAFWVAEVICAASTAAFFLLRNIRTKMGRVSKKAE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias G6PT1/G6PT2/G6PT3/GSD1b/GSD1c/GSD1d/MGC15729/PRO0685/TRG19
Common Name SLC37A4
Gene Symbol SLC37A4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.