Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC39A10 Full-length ORF (NP_065075.1, 1 a.a. - 831 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962328 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-328 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-328 Supplier Abnova™ Supplier No. H00057181P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Zinc is an essential cofactor for hundreds of enzymes. It is involved in protein, nucleic acid, carbohydrate, and lipid metabolism, as well as in the control of gene transcription, growth, development, and differentiation. SLC39A10 belongs to a subfamily of proteins that show structural characteristics of zinc transporters (Taylor and Nicholson, 2003 [PubMed 12659941]).[supplied by OMIM]

Sequence: MKVHMHTKFCLICLLTFIFHHCNHCHEEHDHGPEALHRQHRGMTELEPSKFSKQAAENEKKYYIEKLFERYGENGRLSFFGLEKLLTNLGLGERKVVEINHEDLGHDHVSHLDILAVQEGKHFHSHNHQHSHNHLNSENQTVTSVSTKRNHKCDPEKETVEVSVKSDDKHMHDHNHRLRHHHRLHHHLDHNNTHHFHNDSITPSERGEPSNEPSTETNKTQEQSDVKLPKGKRKKKGRKSNENSEVITPGFPPNHDQGEQYEHNRVHKPDRVHNPGHSHVHLPERNGHDPGRGHQDLDPDNEGELRHTRKREAPHVKNNAIISLRKDLNEDDHHHECLNVTQLLKYYGHGANSPISTDLFTYLCPALLYQIDSRLCIEHFDKLLVEDINKDKNLVPEDEANIGASAWICGIISITVISLLSLLGVILVPIINQGCFKFLLTFLVALAVGTMSGDALLHLLPHSQGGHDHSHQHAHGHGHSHGHESNKFLEEYDAVLKGLVALGGIYLLFIIEHCIRMFKHYKQQRGKQKWFMKQNTEESTIGRKLSDHKLNNTPDSDWLQLKPLAGTDDSVVSEDRLNETELTDLEGQQESPPKNYLCIEEEKIIDHSHSDGLHTIHEHDLHAAAHNHHGENKTVLRKHNHQWHHKHSHHSHGPCHSGSDLKETGIANIAWMVIMGDGIHNFSDGLAIGAAFSAGLTGGISTSIAVFCHELPHELGDFAVLLKAGMTVKQAIVYNLLSAMMAYIGMLIGTAVGQYANNITLWIFAVTAGMFLYVALVDMLPEMLHGDGDNEEHGFCPVGQFILQNLGLLFGFAIMLVIALYEDKIVFDIQF

Specifications

Accession Number NP_065075.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57181
Molecular Weight (g/mol) 120.5kDa
Name SLC39A10 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MKVHMHTKFCLICLLTFIFHHCNHCHEEHDHGPEALHRQHRGMTELEPSKFSKQAAENEKKYYIEKLFERYGENGRLSFFGLEKLLTNLGLGERKVVEINHEDLGHDHVSHLDILAVQEGKHFHSHNHQHSHNHLNSENQTVTSVSTKRNHKCDPEKETVEVSVKSDDKHMHDHNHRLRHHHRLHHHLDHNNTHHFHNDSITPSERGEPSNEPSTETNKTQEQSDVKLPKGKRKKKGRKSNENSEVITPGFPPNHDQGEQYEHNRVHKPDRVHNPGHSHVHLPERNGHDPGRGHQDLDPDNEGELRHTRKREAPHVKNNAIISLRKDLNEDDHHHECLNVTQLLKYYGHGANSPISTDLFTYLCPALLYQIDSRLCIEHFDKLLVEDINKDKNLVPEDEANIGASAWICGIISITVISLLSLLGVILVPIINQGCFKFLLTFLVALAVGTMSGDALLHLLPHSQGGHDHSHQHAHGHGHSHGHESNKFLEEYDAVLKGLVALGGIYLLFIIEHCIRMFKHYKQQRGKQKWFMKQNTEESTIGRKLSDHKLNNTPDSDWLQLKPLAGTDDSVVSEDRLNETELTDLEGQQESPPKNYLCIEEEKIIDHSHSDGLHTIHEHDLHAAAHNHHGENKTVLRKHNHQWHHKHSHHSHGPCHSGSDLKETGIANIAWMVIMGDGIHNFSDGLAIGAAFSAGLTGGISTSIAVFCHELPHELGDFAVLLKAGMTVKQAIVYNLLSAMMAYIGMLIGTAVGQYANNITLWIFAVTAGMFLYVALVDMLPEMLHGDGDNEEHGFCPVGQFILQNLGLLFGFAIMLVIALYEDKIVFDIQF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp781L10106/LZT-Hs2/MGC126565/MGC138428
Common Name SLC39A10
Gene Symbol SLC39A10
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.