Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SLC39A9 Synthetic Peptide

Catalog No. PEP1188
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1188 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1188 Supplier Invitrogen™ Supplier No. PEP1188
Add to Cart
Add to Cart

Synthetic Peptide

ZIP9 Synthetic Peptide, PEP-1188, from Invitrogen.

The zinc transporter ZIP9, also known as SLC39A9, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP9 is a multipass membrane protein that belongs to the ZIP transporter subfamily I. It is located at the trans-Golgi network regardless of zinc presence and is thought to be a zinc homeostasis regulator acting in the secretory pathway without significantly altering cytosolic zinc homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 55334
Species Human
Purification Method purified
Accession Number Q9NUM3
Common Name Human SLC39A9
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol SLC39A9
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.