Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC43A1 Full-length ORF (NP_003618.1, 1 a.a. - 559 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962354 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-354 25 μg
89-962-353 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-354 Supplier Abnova™ Supplier No. H00008501P01L

Used for AP, Array, ELISA, WB-Re

SLC43A1 belongs to the system L family of plasma membrane carrier proteins that transports large neutral amino acids (Babu et al., 2003 [PubMed 12930836]).[supplied by OMIM]

Sequence: MAPTLQQAYRRRWWMACTAVLENLFFSAVLLGWGSLLIILKNEGFYSSTCPAESSTNTTQDEQRRWPGCDQQDEMLNLGFTIGSFVLSATTLPLGILMDRFGPRPVRLVGSACFTASCTLMALASRDVEALSPLIFLALSLNGFGGICLTFTSLTLPNMFGNLRSTLMALMIGSYASSAITFPGIKLIYDAGVAFVVIMFTWSGLACLIFLNCTLNWPIEAFPAPEEVNYTKKIKLSGLALDHKVTGDLFYTHVTTMGQRLSQKAPSLEDGSDAFMSPQDVRGTSENLPERSVPLRKSLCSPTFLWSLLTMGMTQLRIIFYMAAVNKMLEYLVTGGQEHETNEQQQKVAETVGFYSSVFGAMQLLCLLTCPLIGYIMDWRIKDCVDAPTQGTVLGDARDGVATKSIRPRYCKIQKLTNAISAFTLTNLLLVGFGITCLINNLHLQFVTFVLHTIVRGFFHSACGSLYAAVFPSNHFGTLTGLQSLISAVFALLQQPLFMAMVGPLKGEPFWVNLGLLLFSLLGFLLPSYLFYYRARLQQEYAANGMGPLKVLSGSEVTA

Specifications

Accession Number NP_003618.1
For Use With (Application) Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8501
Molecular Weight (g/mol) 87.9kDa
Name Human SLC43A1 Full-length ORF (NP_003618.1, 1 a.a. - 559 a.a.) Recombinant Protein with GST-tag at N-terminal
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Storage Requirements Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias LAT3, PB39, POV1, R00504
Common Name SLC43A1
Gene Symbol SLC43A1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST Tag
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.