Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SLC45A1 (aa 448-523) Control Fragment Recombinant Protein

Catalog No. RP99823
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99823 Supplier Invitrogen™ Supplier No. RP99823
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140021. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene was isolated initially from a region on chromosome 1p that is frequently deleted in human neuroblastoma, although no causal relationship has since been demonstrated. The encoded protein belongs to the glycoside-pentoside-hexuronide cation symporter transporter family and may play a role in glucose uptake.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y2W3
Concentration 3.90 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 50651
Name Human SLC45A1 (aa 448-523) Control Fragment
pH Range 7.4
Purification Method Metal-chelate chromatography
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias deleted in neuroblastoma 5 protein; DNB5; DNb-5; H+/sugar symporter; PAST-A; Proton-associated sugar transporter A; SLC45A1; Solute carrier family 45 member 1; solute carrier family 45, member 1
Common Name SLC45A1
Gene Symbol SLC45A1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QDRGLLEGREGALTSGCDGDILRVGSLDTSKPRSSGILKRPQTLAIPDAAGGGGPETSRRRNVTFSQQVANILLNG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.